siRNA Details
| Gene Symbol: LOC106699570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133569.1 | 2 | 1264 | |||
| GenBank mRNA | AL080152.1 | - | 2 | 1264 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106699570| Gene Symbol: LOC106699570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133569.1 | 2 | 1264 | |||
| GenBank mRNA | AL080152.1 | - | 2 | 1264 | ||