siRNA Details
| Gene Symbol: LINC00996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034033.1 | 3, 4 | 988 | |||
| GenBank mRNA | AK097158.1 | - | 3, 4 | 890 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00996| Gene Symbol: LINC00996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034033.1 | 3, 4 | 988 | |||
| GenBank mRNA | AK097158.1 | - | 3, 4 | 890 | ||