siRNA Details
| Gene Symbol: RNASEH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038429.1 | 1 | 431 | |||
| NR_038430.1 | 1 | 430 | ||||
| NR_038431.1 | 1 | 430 | ||||
| GenBank mRNA | AK023018.1 | - | 1 | 269 | ||
| BC040057.1 | - | 1 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNASEH1-AS1| Gene Symbol: RNASEH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038429.1 | 1 | 431 | |||
| NR_038430.1 | 1 | 430 | ||||
| NR_038431.1 | 1 | 430 | ||||
| GenBank mRNA | AK023018.1 | - | 1 | 269 | ||
| BC040057.1 | - | 1 | 325 | |||