siRNA Details
Gene Symbol: SLIT2-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037876.2 | 4 | 359 | |||
GenBank mRNA | BC041440.1 | - | 4 | 352 | ||
BI831389.1 | - | 4 | 351 | |||
BX089083.1 | - | 4 | 359 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY036413.1 | - | 4 | 434 | Chr. 4 - 20392151 - 20394644 [+] on Build GRCh38 |
