siRNA Details
| Gene Symbol: TPTEP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094889.1 | - | 1 | 1079 | ||
| AK097082.1 | - | 1 | 1646 | |||
| BC031001.1 | - | 3 | 709 | |||
| EU541207.1 | - | 3 | 655 | |||
| M27826.1 | - | 2 | 182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPTEP1| Gene Symbol: TPTEP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094889.1 | - | 1 | 1079 | ||
| AK097082.1 | - | 1 | 1646 | |||
| BC031001.1 | - | 3 | 709 | |||
| EU541207.1 | - | 3 | 655 | |||
| M27826.1 | - | 2 | 182 | |||