siRNA Details
| Gene Symbol: LOC101928896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120566.1 | 1 | 116 | |||
| GenBank mRNA | BC042472.1 | - | 1 | 116 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG717577.1 | - | 1 | 121 | Chr. 11 - 78533176 - 78558551 [+] on Build GRCh38 | |

LOC101928896
Genomic Map