siRNA Details
| Gene Symbol: MIR17HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027350.1 | 2 | 865 | |||
| GenBank mRNA | BC109081.2 | - | 2 | 743 | ||
| BC109082.2 | - | 3 | 358 | |||
| Gene Symbol: MIR18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AB176708.1 | - | LIT3364 oncomir-1 |
2 | 865 | |

MIR17HG
Genomic Map