siRNA Details
| Gene Symbol: SPATA3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033879.1 | 6 | 3223 | |||
| GenBank mRNA | BC047542.1 | - | 7 | 3197 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA3-AS1| Gene Symbol: SPATA3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033879.1 | 6 | 3223 | |||
| GenBank mRNA | BC047542.1 | - | 7 | 3197 | ||