siRNA Details
| Gene Symbol: LINC01235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033863.1 | 3 | 1964 | |||
| GenBank mRNA | AK123194.1 | - | 3 | 1964 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000604724.5 | - | 3 | 1964 | Chr. 9 - 13406380 - 13431329 [-] on Build GRCh38 | |

LINC01235
Genomic Map