siRNA Details
| Gene Symbol: ZNF436-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033690.1 | 1 | 2781 | |||
| NR_033691.1 | 3 | 606 | ||||
| GenBank mRNA | AK074989.1 | - | 1 | 2696 | ||
| BC028411.1 | - | 1 | 2522 | 4822173 | ||
| BC054115.1 | - | 1 | 2785 | 5270027 | ||
| BC068594.1 | - | 1 | 2763 | 4828713 | ||
| BX098506.1 | - | 3 | 574 | |||

ZNF436-AS1
Genomic Map