siRNA Details
| Gene Symbol: LOC101928881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136325.1 | 3 | 547 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000409905.1 | - | 3 | 547 | Chr. 2 - 233012614 - 233015885 [+] on Build GRCh38 | |

LOC101928881
Genomic Map