siRNA Details
| Gene Symbol: LOC102724312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125994.1 | 2 | 437 | |||
| GenBank mRNA | BM726062.1 | - | 2 | 384 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417917.1 | - | 2 | 384 | Chr. 1 - 1430664 - 1434520 [-] on Build GRCh38 | |

LOC102724312
Genomic Map