siRNA Details
| Gene Symbol: LINC01559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036555.1 | 1, 2 | 237 | |||
| GenBank mRNA | AK091129.1 | - | 1, 2 | 203 | ||
| BC101220.1 | - | 1, 2 | 54 | 40022214 | ||
| BC101222.2 | - | 1, 2 | 54 | 40022216 | ||
| BC101223.1 | - | 1, 2 | 54 | 40022217 | ||
| BG421494.1 | - | 238 | ||||

LINC01559
Genomic Map