siRNA Details
| Gene Symbol: LOC284912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC001801.1 | - | 2 | 707 | 3355513 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000472240.1 | - | 2 | 707 | Chr. 22 - 35626988 - 35635134 [-] on Build GRCh38 | |

LOC284912
Genomic Map