siRNA Details
| Gene Symbol: IFNG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104124.1 | 1 | 98 | |||
| NR_104125.1 | 1 | 98 | ||||
| GenBank mRNA | AK124066.1 | - | 1 | 98 | ||
| BG756823.1 | - | 1 | 87 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFNG-AS1| Gene Symbol: IFNG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104124.1 | 1 | 98 | |||
| NR_104125.1 | 1 | 98 | ||||
| GenBank mRNA | AK124066.1 | - | 1 | 98 | ||
| BG756823.1 | - | 1 | 87 | |||