siRNA Details
| Gene Symbol: LINC01234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110025.1 | 2 | 1762 | |||
| NR_110026.1 | 3 | 1896 | ||||
| GenBank mRNA | AK096932.1 | - | 2 | 1761 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000547963.1 | - | 2 | 1762 | Chr. 12 - 113744577 - 113773683 [-] on Build GRCh38 | |

LINC01234
Genomic Map