siRNA Details
| Gene Symbol: KCNQ5-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120503.1 | 1, 2 | 98 | |||
| GenBank mRNA | BX104605.1 | - | 1, 2 | 98 | ||
| HY037383.1 | - | 1, 2 | 239 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000445310.1 | - | 1, 2 | 98 | Chr. 6 - 72630495 - 72678558 [+] on Build GRCh38 | |

KCNQ5-IT1
Genomic Map