siRNA Details
| Gene Symbol: NKX2-1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103710.1 | 1 | 352 | |||
| GenBank mRNA | BX161496.1 | - | 1 | 314 | ||
| DA256249.1 | - | 1 | 352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NKX2-1-AS1| Gene Symbol: NKX2-1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103710.1 | 1 | 352 | |||
| GenBank mRNA | BX161496.1 | - | 1 | 314 | ||
| DA256249.1 | - | 1 | 352 | |||