siRNA Details
| Gene Symbol: LINC01486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120462.1 | 2 | 339 | |||
| GenBank mRNA | BX107914.1 | - | 2 | 339 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000538041.1 | - | 2 | 339 | Chr. 12 - 109354083 - 109359488 [-] on Build GRCh38 | |

LINC01486
Genomic Map