siRNA Details
| Gene Symbol: LOC339059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120387.1 | 3 | 2053 | |||
| GenBank mRNA | AK097767.1 | - | 3 | 2056 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000333666.1 | - | 3 | 2053 | Chr. 16 - 88742767 - 88745748 [+] on Build GRCh38 | |

LOC339059
Genomic Map