siRNA Details
| Gene Symbol: LINC00673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036488.1 | 2, 3 | 700 | |||
| NR_137281.1 | 2, 3 | 702 | ||||
| GenBank mRNA | AK000477.1 | - | 2, 3 | 700 | ||
| AK129763.1 | - | 3, 4 | 435 | |||
| KR778895.1 | - | 2, 3 | 700 | |||

LINC00673
Genomic Map