siRNA Details
Gene Symbol: MYCNOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026766.1 | 1 | 93 | |||
NR_110230.1 | 1 | 93 | ||||
GenBank mRNA | S49953.1 | - | 1 | 93 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000419083.5 | - | 1 | 93 | Chr. 2 - 15939898 - 15941723 [-] on Build GRCh38 |
