siRNA Details
| Gene Symbol: LOC101929231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110240.1 | 2 | 512 | |||
| GenBank mRNA | BM723461.1 | - | 2 | 512 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000449714.1 | - | 2 | 674 | Chr. 2 - 149587196 - 149848233 [+] on Build GRCh38 | |

LOC101929231
Genomic Map