siRNA Details
| Gene Symbol: LOC101929243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134625.1 | 2 | 791 | |||
| NR_134626.1 | 2 | 275 | ||||
| GenBank mRNA | CB045917.1 | - | 2 | 284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929243| Gene Symbol: LOC101929243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134625.1 | 2 | 791 | |||
| NR_134626.1 | 2 | 275 | ||||
| GenBank mRNA | CB045917.1 | - | 2 | 284 | ||