siRNA Details
| Gene Symbol: FIRRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026975.1 | 10 | 1801 | |||
| GenBank mRNA | AK097911.1 | - | 10 | 1800 | ||
| BC131554.1 | - | 10 | 1257 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FIRRE| Gene Symbol: FIRRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026975.1 | 10 | 1801 | |||
| GenBank mRNA | AK097911.1 | - | 10 | 1800 | ||
| BC131554.1 | - | 10 | 1257 | |||