siRNA Details
| Gene Symbol: 2010002M12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053217.2 | NP_444447.1 | 3 | 956 | ||
| GenBank mRNA | AK008052.1 | - | 2 | 808 | ||
| AK136525.1 | - | 2 | 808 | |||
| AK153247.1 | - | 3 | 957 | |||
| BC003493.1 | - | 1 | 309 | |||
| BC138202.1 | - | 2 | 787 | 8861221 | ||
 Back To Top
 Back To TopMore Information
Other Species Targeted
 This siRNA also targets the following transcripts in Rat: XM_008760355, XM_220058.
					            	This siRNA also targets the following transcripts in Rat: XM_008760355, XM_220058.Gene Ontology Categories:
 Back To Top
 Back To Top
 2010002M12Rik
2010002M12Rik Genomic Map
Genomic Map Back To Top
 Back To Top