siRNA Details
| Gene Symbol: 2010002M12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053217.2 | NP_444447.1 | 3 | 1116 | ||
| GenBank mRNA | AK136525.1 | - | 2 | 968 | ||
| AK153247.1 | - | 3 | 1117 | |||
| BC003493.1 | - | 1 | 470 | |||
| BC138202.1 | - | 2 | 947 | 8861221 | ||

2010002M12Rik
Genomic Map