siRNA Details
| Gene Symbol: Vmn1r41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053230.2 | NP_444460.2 | 2 | 186 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r41| Gene Symbol: Vmn1r41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053230.2 | NP_444460.2 | 2 | 186 | ||