siRNA Details
| Gene Symbol: Vmn1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053231.2 | NP_444461.2 | 1 | 757 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r10| Gene Symbol: Vmn1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053231.2 | NP_444461.2 | 1 | 757 | ||