siRNA Details
| Gene Symbol: Slc28a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022317.3 | NP_071712.3 | 10 | 1073 | ||
| GenBank mRNA | AF305211.1 | - | 10 | 1006 | ||
| AK052784.1 | - | 10 | 1074 | |||
| AK145077.1 | - | 10 | 1074 | |||
| BC010472.1 | - | 10 | 1061 | 3586005 | ||
| BC013783.1 | - | 10 | 1064 | 3600786 | ||

Slc28a3
Genomic Map