siRNA Details
| Gene Symbol: Zbtb21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081684.1 | NP_001075153.1 | 2 | 235 | ||
| NM_001081685.1 | NP_001075154.1 | 3 | 378 | |||
| NM_175428.3 | NP_780637.2 | 3 | 378 | |||
| GenBank mRNA | AK031118.1 | - | 3 | 367 | ||
| AK034057.1 | - | 3 | 379 | |||
| AK129313.1 | - | 2 | 212 | |||
| AK145072.1 | - | 2 | 230 | |||
| BC027135.1 | - | 1 | 32 | 5360683 | ||
| BC051176.1 | - | 2 | 227 | |||

Zbtb21
Genomic Map