siRNA Details
| Gene Symbol: Oas1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033541.4 | NP_291019.1 | 3 | 663 | ||
| GenBank mRNA | AB067528.1 | - | 2 | 320 | ||
| AF459815.1 | - | 2 | 320 | |||
| AF459816.1 | - | 2 | 320 | |||
| AK028986.1 | - | 3 | 664 | |||
| AK078952.1 | - | 2 | 331 | |||
| AK163253.1 | - | 2 | 336 | |||
| BC108955.1 | - | 2 | 329 | 40046017 | ||
| BC126877.1 | - | 2 | 328 | 40046021 | ||

Oas1c
Genomic Map