siRNA Details
| Gene Symbol: Slc13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054055.2 | NP_473396.1 | 10 | 1308 | ||
| GenBank mRNA | AF306491.1 | - | 10 | 1307 | ||
| AK085326.1 | - | 10 | 1306 | |||
| AK128982.1 | - | 10 | 1305 | |||
| AK138344.1 | - | 10 | 1306 | |||
| AK157881.1 | - | 10 | 1277 | |||
| AK165355.1 | - | 10 | 1311 | |||
| BC026803.1 | - | 10 | 1294 | 4238769 | ||
| BC028882.1 | - | 9 | 1167 | |||

Slc13a3
Genomic Map