siRNA Details
| Gene Symbol: Ly6g5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148947.1 | NP_683749.1 | 2 | 163 | ||
| GenBank mRNA | AJ315552.1 | - | 2 | 163 | ||
| BC111447.1 | - | 2 | 162 | |||
| BC145966.1 | - | 2 | 184 | 40131269 | ||
| BC145968.1 | - | 2 | 184 | 40131271 | ||

Ly6g5c
Genomic Map