siRNA Details
| Gene Symbol: Hsd17b11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053262.3 | NP_444492.1 | 5, 6 | 746 | ||
| GenBank mRNA | AF304306.1 | - | 5, 6 | 742 | ||
| AK154803.1 | - | 5, 6 | 761 | |||
| AK155174.1 | - | 5, 6 | 761 | |||
| AK155202.1 | - | 5, 6 | 766 | |||
| AK155327.1 | - | 5, 6 | 766 | |||
| AK158327.1 | - | 5, 6 | 761 | |||
| AK170939.1 | - | 5, 6 | 761 | |||
| AY053570.1 | - | 5, 6 | 760 | |||
| BC038340.1 | - | 5, 6 | 746 | 4458725 | ||

Hsd17b11
Genomic Map