siRNA Details
Gene Symbol: AF366264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153093.3 | NP_694733.3 | 1 | 380 |
Gene Symbol: 4930444G20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053264.2 | NP_444494.1 | 1 | 359 |
