siRNA Details
| Gene Symbol: 4930444G20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053264.2 | NP_444494.1 | 1 | 629 | ||
| GenBank mRNA | AK015380.1 | - | 1 | 216 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930444G20Rik| Gene Symbol: 4930444G20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053264.2 | NP_444494.1 | 1 | 629 | ||
| GenBank mRNA | AK015380.1 | - | 1 | 216 | ||