siRNA Details
Gene Symbol: 4930444G20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053264.2 | NP_444494.1 | 1 | 509 | ||
GenBank mRNA | AK015380.1 | - | 1 | 96 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930444G20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053264.2 | NP_444494.1 | 1 | 509 | ||
GenBank mRNA | AK015380.1 | - | 1 | 96 |