siRNA Details
| Gene Symbol: Selm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053267.2 | NP_444497.1 | 4, 5 | 521 | ||
| GenBank mRNA | AK018791.1 | - | 4, 5 | 357 | ||
| AK035312.1 | - | 4, 5 | 311 | |||
| AK046034.1 | - | 4, 5 | 313 | |||
| AK079358.1 | - | 4, 5 | 340 | |||
| AY043488.1 | - | 4, 5 | 337 | |||
| BC019742.1 | - | 4, 5 | 213 | |||
| BC147332.1 | - | 4, 5 | 431 | 9056406 | ||
| BC147333.1 | - | 4, 5 | 433 | 9056396 | ||
| CK787580.1 | - | 4, 5 | 513 | |||

Selm
Genomic Map