siRNA Details
| Gene Symbol: Psg29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054064.3 | NP_473405.1 | 2 | 546 | ||
| GenBank mRNA | AK141330.1 | - | 2 | 547 | ||
| BC114207.1 | - | 2 | 523 | 30083585 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psg29| Gene Symbol: Psg29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054064.3 | NP_473405.1 | 2 | 546 | ||
| GenBank mRNA | AK141330.1 | - | 2 | 547 | ||
| BC114207.1 | - | 2 | 523 | 30083585 | ||