siRNA Details
| Gene Symbol: Fgfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054071.2 | NP_473412.1 | 3 | 626 | ||
| GenBank mRNA | AF321300.1 | - | 3 | 353 | ||
| AJ293947.1 | - | 2 | 259 | |||
| BC058745.1 | - | 2 | 228 | 6439421 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgfrl1| Gene Symbol: Fgfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054071.2 | NP_473412.1 | 3 | 626 | ||
| GenBank mRNA | AF321300.1 | - | 3 | 353 | ||
| AJ293947.1 | - | 2 | 259 | |||
| BC058745.1 | - | 2 | 228 | 6439421 | ||