siRNA Details
| Gene Symbol: Tsga13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054073.2 | NP_473414.1 | 5 | 528 | ||
| GenBank mRNA | AF294348.1 | - | 5 | 496 | ||
| AK006265.1 | - | 5 | 529 | |||
| BC125597.1 | - | 4 | 446 | 40130112 | ||
| BC125599.1 | - | 4 | 446 | 40130114 | ||

Tsga13
Genomic Map