siRNA Details
| Gene Symbol: Lsm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163266.1 | NP_001156738.1 | 2 | 221 | ||
| NM_138721.2 | NP_620046.1 | 2 | 323 | |||
| GenBank mRNA | AK143443.1 | - | 2 | 262 | ||
| BC024543.1 | - | 2 | 224 | 4989202 | ||
| BC026476.1 | - | 2 | 252 | 4489522 | ||

Lsm10
Genomic Map