siRNA Details
| Gene Symbol: Mta3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171052.1 | NP_001164523.1 | 7 | 619 | ||
| NM_001171053.1 | NP_001164524.1 | 7 | 619 | |||
| NM_001171054.1 | NP_001164525.1 | 7 | 619 | |||
| NM_054082.2 | NP_473423.1 | 7 | 616 | |||
| GenBank mRNA | AF288138.1 | - | 7 | 570 | ||
| AK129318.1 | - | 7 | 548 | |||
| AK145942.1 | - | 7 | 564 | |||
| AK146721.1 | - | 7 | 572 | |||
| AK146905.1 | - | 7 | 572 | |||
| AK147197.1 | - | 7 | 562 | |||
| AK154856.1 | - | 7 | 559 | |||
| AK158866.1 | - | 6 | 482 | |||
| BC022124.1 | - | 7 | 583 | 5097283 | ||
| BC099960.1 | - | 7 | 536 | 30023887 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008764474.

Mta3
Genomic Map