siRNA Details
| Gene Symbol: Derl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033562.4 | NP_291040.1 | 1, 2 | 163 | ||
| GenBank mRNA | AF208064.1 | - | 1, 2 | 102 | ||
| AK048627.1 | - | 1, 2 | 110 | |||
| AK077659.1 | - | 1, 2 | 114 | |||
| AK082564.1 | - | 1, 2 | 123 | |||
| AK083632.1 | - | 1, 2 | 102 | |||
| BC005682.1 | - | 1, 2 | 102 | 3154448 | ||

Derl2
Genomic Map