siRNA Details
| Gene Symbol: Cts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026906.3 | NP_081182.2 | 7 | 979 | ||
| NM_054092.2 | NP_473433.2 | 7 | 904 | |||
| GenBank mRNA | AK005389.1 | - | 7 | 902 | ||
| AY034574.1 | - | 7 | 918 | |||
| BC099388.1 | - | 7 | 873 | 30786828 | ||

Cts3
Genomic Map