siRNA Details
| Gene Symbol: Ube3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054093.2 | NP_473434.2 | 19 | 2468 | ||
| GenBank mRNA | AF244362.3 | - | 18 | 2103 | ||
| AK088518.1 | - | 18 | 2164 | |||
| BC023956.1 | - | 19 | 2237 | 5321098 | ||
| BC034059.1 | - | 13 | 1472 | 4163877 | ||
| BC096743.1 | - | 19 | 2491 | 30549276 | ||

Ube3b
Genomic Map