siRNA Details
| Gene Symbol: Acsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054094.5 | NP_473435.1 | 9 | 1232 | ||
| GenBank mRNA | AB059428.1 | - | 9 | 1236 | ||
| AK149550.1 | - | 9 | 1242 | |||
| BC016414.1 | - | 9 | 1229 | 4238964 | ||
| BC022149.1 | - | 9 | 1210 | 5102652 | ||

Acsm1
Genomic Map