siRNA Details
| Gene Symbol: 2310034C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054100.2 | NP_473441.1 | 1 | 854 | ||
| GenBank mRNA | AK009607.1 | - | 1 | 855 | ||
| BC064816.1 | - | 1 | 836 | 30287188 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310034C09Rik| Gene Symbol: 2310034C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054100.2 | NP_473441.1 | 1 | 854 | ||
| GenBank mRNA | AK009607.1 | - | 1 | 855 | ||
| BC064816.1 | - | 1 | 836 | 30287188 | ||