siRNA Details
| Gene Symbol: Bloc1s4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133724.3 | NP_598485.1 | 1 | 1078 | ||
| GenBank mRNA | AK011795.1 | - | 1 | 1050 | ||
| AK182170.1 | - | 1 | 191 | |||
| AK207410.1 | - | 1 | 311 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bloc1s4| Gene Symbol: Bloc1s4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133724.3 | NP_598485.1 | 1 | 1078 | ||
| GenBank mRNA | AK011795.1 | - | 1 | 1050 | ||
| AK182170.1 | - | 1 | 191 | |||
| AK207410.1 | - | 1 | 311 | |||